missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Chlorophenylethynyl)trimethylsilane, 97%

$31.49 - $491.80

Chemical Identifiers

CAS 78704-49-1
Molecular Formula C11H13ClSi
Molecular Weight (g/mol) 208.76
MDL Number MFCD03427267
InChI Key HVIHXUCWKNRJTC-UHFFFAOYSA-N
Synonym 2 4'-chlorophenyl-1-trimethylsilylethyne
PubChem CID 3337298
IUPAC Name 2-(4-chlorophenyl)ethynyl-trimethylsilane
SMILES C[Si](C)(C)C#CC1=CC=C(C=C1)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5512903 View Documents Alfa Aesar™
H5512903
1g Each for $31.49
 
AAH5512906 View Documents Alfa Aesar™
H5512906
5g Each for $109.62
 
AAH5512914 View Documents Alfa Aesar™
H5512914
25g Each for $491.80
 
Specifications

Chemical Identifiers

78704-49-1
208.76
HVIHXUCWKNRJTC-UHFFFAOYSA-N
3337298
C[Si](C)(C)C#CC1=CC=C(C=C1)Cl
C11H13ClSi
MFCD03427267
2 4'-chlorophenyl-1-trimethylsilylethyne
2-(4-chlorophenyl)ethynyl-trimethylsilane

Specifications

78704-49-1
MFCD03427267
HVIHXUCWKNRJTC-UHFFFAOYSA-N
2-(4-chlorophenyl)ethynyl-trimethylsilane
3337298
Solid
Brown to White
1g
(4-Chlorophenylethynyl)trimethylsilane
C11H13ClSi
2 4'-chlorophenyl-1-trimethylsilylethyne
C[Si](C)(C)C#CC1=CC=C(C=C1)Cl
208.76
208.76
97%
47°C to 51°C
104°C (219°F)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures