missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Chlorophenylethynyl)trimethylsilane, 97%

Manufacturer:  Alfa Aesar™ H5512906

 View more versions of this product

Catalog No. AAH5512906



Specifications

Specifications

(4-Chlorophenylethynyl)trimethylsilane
C11H13ClSi
2 4'-chlorophenyl-1-trimethylsilylethyne
C[Si](C)(C)C#CC1=CC=C(C=C1)Cl
208.76
208.76
97%
47°C to 51°C
104°C (219°F)
78704-49-1
MFCD03427267
HVIHXUCWKNRJTC-UHFFFAOYSA-N
2-(4-chlorophenyl)ethynyl-trimethylsilane
3337298
Solid
Brown to White
5g

Chemical Identifiers

78704-49-1
208.76
HVIHXUCWKNRJTC-UHFFFAOYSA-N
3337298
C[Si](C)(C)C#CC1=CC=C(C=C1)Cl
C11H13ClSi
MFCD03427267
2 4'-chlorophenyl-1-trimethylsilylethyne
2-(4-chlorophenyl)ethynyl-trimethylsilane
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents