missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorophenylboronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444690250

 View more versions of this product

Catalog No. AC444690250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Chlorophenylboronic acid pinacol ester
55.0°C to 56.0°C
195062-61-4
C12H16BClO2
NYARTXMDWRAVIX-UHFFFAOYSA-N
2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
10633712
97%
>110°C
25g
96% min. (GC)
1,3,2-dioxaborolane, 2-4-chlorophenyl-4,4,5,5-tetramethyl, 1,3,2-dioxaborolane,2-4-chlorophenyl-4,4,5,5-tetramethyl, 2-4-chlorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chlorobenzene, 4-chlorobenzeneboronic acid, pinacol ester, 4-chlorophenyl boronic acid pinacol ester, 4-chlorophenylboronic acid pinacol ester, 4-chlorophenylboronic acid, pinacol ester, p-chlorophenylboronic acid 2,3-dimethyl-2,3-butanediyl
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)Cl
238.52
238.52

Chemical Identifiers

195062-61-4
238.52
1,3,2-dioxaborolane, 2-4-chlorophenyl-4,4,5,5-tetramethyl, 1,3,2-dioxaborolane,2-4-chlorophenyl-4,4,5,5-tetramethyl, 2-4-chlorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chlorobenzene, 4-chlorobenzeneboronic acid, pinacol ester, 4-chlorophenyl boronic acid pinacol ester, 4-chlorophenylboronic acid pinacol ester, 4-chlorophenylboronic acid, pinacol ester, p-chlorophenylboronic acid 2,3-dimethyl-2,3-butanediyl
2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C12H16BClO2
NYARTXMDWRAVIX-UHFFFAOYSA-N
10633712
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)Cl
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents