missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorophenylboronic acid, 97%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 344410050

 View more versions of this product

Catalog No. AC344410050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Chlorophenylboronic acid
Glass bottle
259.0°C to 262.0°C
1679-18-1
100.0
C6H6BClO2
4-chlorobenzeneboronic acid, 4-chlorophenyl boranediol, 4-chlorophenyl boronic acid, 4-chlorophenylbornic acid, benzeneboronic acid, p-chloro, boronic acid, 4-chlorophenyl, boronic acid, p-chlorophenyl, p-chlorobenzeneboronic acid, p-chlorophenylboronic acid
CAYQIZIAYYNFCS-UHFFFAOYSA-N
(4-chlorophenyl)boronic acid
74299
Crystalline Powder
Authentic
White to Beige
5g
96.0
96% min. (HPLC)
ClC6H4B(OH)2
Solubility in water:+/- 2.5% in water
B(C1=CC=C(C=C1)Cl)(O)O
156.38
156.38
97%

Chemical Identifiers

1679-18-1
156.38
4-chlorobenzeneboronic acid, 4-chlorophenyl boranediol, 4-chlorophenyl boronic acid, 4-chlorophenylbornic acid, benzeneboronic acid, p-chloro, boronic acid, 4-chlorophenyl, boronic acid, p-chlorophenyl, p-chlorobenzeneboronic acid, p-chlorophenylboronic acid
(4-chlorophenyl)boronic acid
C6H6BClO2
CAYQIZIAYYNFCS-UHFFFAOYSA-N
74299
B(C1=CC=C(C=C1)Cl)(O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-845-5

SDS Documents