Learn More
4-Chlorophenylacetylene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5105803
Description
4-Chlorophenylacetylene is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Chlorophenylacetylene is used as pharmaceutical intermediate.
Solubility
Insoluble in water. Soluble in acetone, chloroform, dichloromethane, DMF, DMSO, ethanol, ethyl acetate, hexane, methanol,THF and toluene.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
4-Chlorophenylacetylene | |
873-73-4 | |
1 g | |
MFCD00191917 | |
4-chlorophenylacetylene, benzene, 1-chloro-4-ethynyl, 4'-chlorophenyl acetylene, 4-chlorophenyl acetylene, 1-chloro-4-ethynyl-benzene, p-ethynylchlorobenzene, p-chlorophenylacetylene, p-chlorophenyl acetylene, 1-chlor-4-ethinylbenzol, 4-chlorophenylethyne | |
LFZJRTMTKGYJRS-UHFFFAOYSA-N | |
1-chloro-4-ethynylbenzene | |
70118 | |
98% |
43°C to 47°C | |
179°C | |
C8H5Cl | |
UN1325 | |
Insoluble in water. Soluble in acetone,chloroform,dichloromethane,DMF,DMSO,ethanol,ethyl acetate,hexane,methanol,THF and toluene. | |
C#CC1=CC=C(C=C1)Cl | |
136.578 | |
136.58 |
Chemical Identifiers
873-73-4 | |
136.578 | |
LFZJRTMTKGYJRS-UHFFFAOYSA-N | |
70118 | |
C#CC1=CC=C(C=C1)Cl |
C8H5Cl | |
MFCD00191917 | |
4-chlorophenylacetylene, benzene, 1-chloro-4-ethynyl, 4'-chlorophenyl acetylene, 4-chlorophenyl acetylene, 1-chloro-4-ethynyl-benzene, p-ethynylchlorobenzene, p-chlorophenylacetylene, p-chlorophenyl acetylene, 1-chlor-4-ethinylbenzol, 4-chlorophenylethyne | |
1-chloro-4-ethynylbenzene |
Safety and Handling
GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Hazard Class: 4.1; Packaging Group: II
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only