missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorophenylacetonitrile, 98+%

Manufacturer:  Alfa Aesar™ A1529536

 View more versions of this product

Catalog No. AAA1529536



Specifications

Specifications

4-Chlorophenylacetonitrile
Pungent
28°C to 31°C
140-53-4
MFCD00001918
971171
IVYMIRMKXZAHRV-UHFFFAOYSA-N
2-(4-chlorophenyl)acetonitrile
241582
151.6
134°C (273°F)
141°C to 143°C (11mmHg)
500g
C8H6ClN
UN3439
4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide
C1=CC(=CC=C1CC#N)Cl
151.593
CHEBI:17346
≥98%

Chemical Identifiers

140-53-4
151.593
IVYMIRMKXZAHRV-UHFFFAOYSA-N
241582
2-(4-chlorophenyl)acetonitrile
C8H6ClN
MFCD00001918
4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide
CHEBI:17346
C1=CC(=CC=C1CC#N)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H300-H311-H331-H315-H319-H335
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

RTECSNumber : AL8250000

TSCA : TSCA

Documents