Please login to your online account to display your discounted pricing

4-chlorophenylacetone, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC350700250

 View more versions of this product

Catalog No. AC350700250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 168.62
Physical Form Liquid
Assay ≥97.5%
Quantity 25g
Color Clear light yellow
Density 1.151g/mL
Beilstein 07, IV, 691
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 4-chlorophenylacetone
Assay Percent Range 98%
CAS 5586-88-9
Molecular Formula C9H9ClO
CAS Min % 97.5
MDL Number MFCD00045214
Refractive Index 1.5325 to 1.5345 (20°C, 589nm)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-chlorophenylacetone
CAS 5586-88-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9ClO
Formula Weight 168.62
MDL Number MFCD00045214