Please login to your online account to display your discounted pricing

4-chlorophenylacetone, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-chlorophenylacetone
CAS 5586-88-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9ClO
Formula Weight 168.62
MDL Number MFCD00045214
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC350700250

  SDS
thermo fisher scientific
AC350700250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC350700050

  SDS
fisher scientific acros
350700050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.62
Physical Form Liquid
Assay ≥97.5%
Quantity 25g
Color Clear light yellow
Density 1.151g/mL
Beilstein 07, IV, 691
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 4-chlorophenylacetone
Assay Percent Range 98%
CAS 5586-88-9
Molecular Formula C9H9ClO
CAS Min % 97.5
MDL Number MFCD00045214
Refractive Index 1.5325 to 1.5345 (20°C, 589nm)
Infrared Spectrum Authentic