missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorophenylacetic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC160720050

 View more versions of this product

Catalog No. AC160720050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(5% soln. in CHCl3) clear
Authentic
0.2% max. (K.F.)
103°C to 107°C
98%
97.5
97.5% min. (HPLC)
ClC6H4CH2CO2H
09,448
Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,diethyl ether,solubility 5% in chcl3: clear solution
C1=CC(=CC=C1CC(=O)O)Cl
170.6
CHEBI:30749
Powder
4-Chlorophenylacetic acid
Glass bottle
White to Cream
5g
1878-66-6
100.0
C8H7ClO2
MFCD00004344
2-4-chlorophenyl acetic acid
CDPKJZJVTHSESZ-UHFFFAOYSA-N
2-(4-chlorophenyl)acetic acid
15880
170.6
98%

Chemical Identifiers

1878-66-6
170.6
CDPKJZJVTHSESZ-UHFFFAOYSA-N
15880
2-(4-chlorophenyl)acetic acid
C8H7ClO2
MFCD00004344
2-4-chlorophenyl acetic acid
CHEBI:30749
C1=CC(=CC=C1CC(=O)O)Cl
Safety and Handling

Safety and Handling

EINECSNumber : 217-521-6

RTECSNumber : AG0590000

TSCA : TSCA

SDS Documents