missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorophenylacetic acid, 98%

Manufacturer:  Alfa Aesar™ A1497430

 View more versions of this product

Catalog No. AAA1497430



Specifications

Specifications

4-Chlorophenylacetic acid
103°C to 107°C
1878-66-6
MFCD00004344
4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl, 4-chlorophenyl acetic acid
C1=CC(=CC=C1CC(=O)O)Cl
170.592
CHEBI:30749
98%
Odorless
250g
C8H7ClO2
1072816
CDPKJZJVTHSESZ-UHFFFAOYSA-N
2-(4-chlorophenyl)acetic acid
15880
170.6

Chemical Identifiers

1878-66-6
170.592
CDPKJZJVTHSESZ-UHFFFAOYSA-N
15880
2-(4-chlorophenyl)acetic acid
C8H7ClO2
MFCD00004344
4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl, 4-chlorophenyl acetic acid
CHEBI:30749
C1=CC(=CC=C1CC(=O)O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : AG0590000

TSCA : TSCA

Documents