Learn More
4-Chlorophenylacetic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1497418
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Chlorophenylacetic acid | |
1878-66-6 | |
Odorless | |
MFCD00004344 | |
4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl | |
OC(=O)CC1=CC=C(Cl)C=C1 | |
170.59 | |
CHEBI:30749 | |
98% |
103°C to 107°C | |
50 g | |
C8H7ClO2 | |
1072816 | |
CDPKJZJVTHSESZ-UHFFFAOYSA-N | |
2-(4-chlorophenyl)acetic acid | |
15880 | |
170.6 |
Chemical Identifiers
1878-66-6 | |
170.59 | |
CDPKJZJVTHSESZ-UHFFFAOYSA-N | |
15880 | |
2-(4-chlorophenyl)acetic acid |
C8H7ClO2 | |
MFCD00004344 | |
4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl | |
CHEBI:30749 | |
OC(=O)CC1=CC=C(Cl)C=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning
EINECSNumber : 217-521-6
RTECSNumber : AG0590000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only