missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorophenyl phosphorodichloridate, 98+%

Manufacturer:  Alfa Aesar™ A1550722

 View more versions of this product

Catalog No. AAA1550722



Description

Description

Applications
4-Chlorophenyl phosphorodichloridate is used as phosphorylation agent. Reactant for preparation of antimicrobial agents, antioxidant agents, antitumor agents, nucleotide anti-viral prodrug.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents, alcohols, bases.
Specifications

Specifications

4-Chlorophenyl phosphorodichloridate
Acrid
100g
C6H4Cl3O2P
UN3265
4-chlorophenyl phosphorodichloridate, p-chlorophenyl dichlorophosphate, 4-chlorophenyl dichlorophosphate, 4-chlorophenylphosphorodichloridate, phosphorodichloridic acid, 4-chlorophenyl ester, 4-chlorophenylphosphoryl dichloride, acmc-1bjgo, cczmqygsxwzfki-uhfffaoysa, 4-chlorophenyl dichlorophosphinate, 4-chlorophenyl dichloridophosphate #
C1=CC(=CC=C1OP(=O)(Cl)Cl)Cl
245.42
245.43
106°C (222°F)
113°C to 115°C (0.9mmHg)
772-79-2
MFCD00009705
910754
CCZMQYGSXWZFKI-UHFFFAOYSA-N
1-chloro-4-dichlorophosphoryloxybenzene
69879
≥98%

Chemical Identifiers

772-79-2
245.42
CCZMQYGSXWZFKI-UHFFFAOYSA-N
69879
C1=CC(=CC=C1OP(=O)(Cl)Cl)Cl
C6H4Cl3O2P
MFCD00009705
4-chlorophenyl phosphorodichloridate, p-chlorophenyl dichlorophosphate, 4-chlorophenyl dichlorophosphate, 4-chlorophenylphosphorodichloridate, phosphorodichloridic acid, 4-chlorophenyl ester, 4-chlorophenylphosphoryl dichloride, acmc-1bjgo, cczmqygsxwzfki-uhfffaoysa, 4-chlorophenyl dichlorophosphinate, 4-chlorophenyl dichloridophosphate #
1-chloro-4-dichlorophosphoryloxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Documents