missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorophenyl methyl sulfone, 98%, ACROS Organics™

Chemical Identifiers

CAS 98-57-7
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H7ClO2S
Formula Weight 190.65
MDL Number MFCD00025067
Chemical Name or Material 4-Chlorophenyl methyl sulfone, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC352200050 SDS Fisher Scientific Acros
352200050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC352200250 SDS Thermo Fisher Scientific
352200250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

98-57-7
100.0
C7H7ClO2S
MFCD00025067
≥97.5%
97.5
98%
190.65
4-Chlorophenyl methyl sulfone, 98%

Specifications

Crystalline Powder
5g
96° to 100°C
C7H7ClO2S
MFCD00025067
97.5
Glass bottle
≥97.5%
190.65
Off-white
100.0
98%
98-57-7
Authentic
4-Chlorophenyl methyl sulfone, 98%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-682-7

RTECSNumber : WR5730000

TSCA : TSCA