missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorophenyl methyl sulfone, 98+%

Manufacturer:  Alfa Aesar™ A1542930

 View more versions of this product

Catalog No. AAA1542930



Specifications

Specifications

4-Chlorophenyl methyl sulfone
250g
C7H7ClO2S
638281
LMCOQDVJBWVNNI-UHFFFAOYSA-N
1-chloro-4-methylsulfonylbenzene
7395
≥98%
95°C to 99°C
98-57-7
MFCD00025067
4-chlorophenyl methyl sulfone, 1-chloro-4-methylsulfonyl benzene, p-chlorophenyl methyl sulfone, 4-methylsulfuryl chlorobenzene, benzene, 1-chloro-4-methylsulfonyl, methyl 4-chlorophenyl sulfone, sulfone, p-chlorophenyl methyl, 1-chloro-4-methylsulphonyl benzene, 4-chlorophenylmethyl sulfone, methyl p-chlorophenyl sulfone
CS(=O)(=O)C1=CC=C(C=C1)Cl
190.641
190.66

Chemical Identifiers

98-57-7
190.641
LMCOQDVJBWVNNI-UHFFFAOYSA-N
7395
CS(=O)(=O)C1=CC=C(C=C1)Cl
C7H7ClO2S
MFCD00025067
4-chlorophenyl methyl sulfone, 1-chloro-4-methylsulfonyl benzene, p-chlorophenyl methyl sulfone, 4-methylsulfuryl chlorobenzene, benzene, 1-chloro-4-methylsulfonyl, methyl 4-chlorophenyl sulfone, sulfone, p-chlorophenyl methyl, 1-chloro-4-methylsulphonyl benzene, 4-chlorophenylmethyl sulfone, methyl p-chlorophenyl sulfone
1-chloro-4-methylsulfonylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : WR5730000

TSCA : TSCA

Documents