Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorophenyl methyl sulfone, 98+%

$80.61 - $295.32

Chemical Identifiers

Chemical Name or Material 4-Chlorophenyl methyl sulfone
CAS 98-57-7
Molecular Formula C7H7ClO2S
Formula Weight 190.66
MDL Number MFCD00025067
Beilstein 638281
Synonym 1-Chloro-4-(methanesulfonyl)benzene; 4-Methanesulfonylchlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1542909

  SDS
alfa aesar™
A1542909
10g Each for $80.61

AAA1542918

  SDS
alfa aesar™
A1542918
50g Each for $295.32
Description & Specifications

Specifications

Formula Weight 190.66
Chemical Name or Material 4-Chlorophenyl methyl sulfone
TSCA TSCA
Assay ≥98%
Molecular Formula C7H7ClO2S
MDL Number MFCD00025067
CAS 98-57-7
Beilstein 638281
Synonym 1-Chloro-4-(methanesulfonyl)benzene; 4-Methanesulfonylchlorobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number WR5730000
Melting Point 95° to 99°C
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.