Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Chlorophenyl)methoxydimethylsilane, 96%

$93.43 - $277.07

Chemical Identifiers

Chemical Name or Material (4-Chlorophenyl)methoxydimethylsilane
CAS 62244-44-4
Molecular Formula C9H13ClOSi
Formula Weight 200.74
MDL Number MFCD09266143
Synonym (4-Chlorophenyl)dimethylmethoxysilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2781003

 
alfa aesar™
H2781003
1g Each for $93.43

AAH2781006

 
alfa aesar™
H2781006
5g Each for $277.07
Description & Specifications

Specifications

Formula Weight 200.74
Chemical Name or Material (4-Chlorophenyl)methoxydimethylsilane
Assay 96%
Molecular Formula C9H13ClOSi
MDL Number MFCD09266143
CAS 62244-44-4
Synonym (4-Chlorophenyl)dimethylmethoxysilane
Boiling Point 93° to 95°C (15mmHg)
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.