Please login to your online account to display your discounted pricing

4-Chlorophenyl isocyanate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chlorophenyl isocyanate
CAS 104-12-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4ClNO
Linear Formula ClC6H4NCO
Formula Weight 153.57
MDL Number MFCD00002024
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC166565000

  SDS
thermo fisher scientific
AC166565000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC166560250

  SDS
thermo fisher scientific
166560250
Glass Bottle in Tin Can 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.57
Assay ≥97.5%
Physical Form Crystalline Solid
Quantity 500g
Color White to yellow
Melting Point 29° to 31°C
Density 1.2600g/mL
Beilstein 12, 616
CAS Max % 100.0
Chemical Name or Material 4-Chlorophenyl isocyanate
CAS 104-12-1
Packaging Glass bottle
Molecular Formula C7H4ClNO
Assay Percent Range 98%
MDL Number MFCD00002024
CAS Min % 97.5
Infrared Spectrum Authentic
Storage Information Product may develop some turbidity or precipitate
Linear Formula ClC6H4NCO
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Fatal if inhaled.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Fatal if inhaled.
Toxic if swallowed.