Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorophenyl cyclopropyl ketone, 98%

$60.66 - $868.15

Chemical Identifiers

Chemical Name or Material 4-Chlorophenyl cyclopropyl ketone
CAS 6640-25-1
Molecular Formula C10H9ClO
Formula Weight 180.63
MDL Number MFCD00001295
Beilstein 2044845
Synonym (4-Chlorobenzoyl)cyclopropane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2276609

 
alfa aesar™
B2276609
10g Each for $60.66

AAB2276618

 
alfa aesar™
B2276618
50g Each for $218.58

AAB2276630

 
alfa aesar™
B2276630
250g Each for $868.15
Description & Specifications

Specifications

Formula Weight 180.63
Chemical Name or Material 4-Chlorophenyl cyclopropyl ketone
Assay 98%
Molecular Formula C10H9ClO
MDL Number MFCD00001295
CAS 6640-25-1
Beilstein 2044845
Synonym (4-Chlorobenzoyl)cyclopropane
Flash Point >110°C (230°F)
Boiling Point 135° to 137°C (10mmHg)
Melting Point 29° to 31°C
Quantity 10g