missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorophenoxyacetyl Chloride 98.0+%, TCI America™

$137.70 - $1093.29

Chemical Identifiers

CAS 4122-68-3
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00000727
InChI Key VRBVHQUSAOKVDH-UHFFFAOYSA-N
Synonym 4-chlorophenoxyacetyl chloride, p-chlorophenoxyacetyl chloride, 2-4-chlorophenoxy acetyl chloride, 4-chlorophenoxy acetyl chloride, acetyl chloride, 4-chlorophenoxy, 4-chlorophenoxyacetylchloride, acmc-1aqz7, 4-chlorophenoxyacetyl chlroride, 4chlorophenoxy acetyl chloride, vrbvhqusaokvdh-uhfffaoysa
PubChem CID 77771
IUPAC Name 2-(4-chlorophenoxy)acetyl chloride
SMILES C1=CC(=CC=C1OCC(=O)Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C083225G
View Documents Promotion Details
TCI America
C083225G
25 g
Each for $137.70
 
C0832100G
View Documents Promotion Details
TCI America
C0832100G
100 g
Each for $378.25
 
C0832500G
View Documents Promotion Details
TCI America
C0832500G
500 g
Each for $1,093.29
 
Description

Description

Specifications

Chemical Identifiers

4122-68-3
205.034
VRBVHQUSAOKVDH-UHFFFAOYSA-N
77771
C1=CC(=CC=C1OCC(=O)Cl)Cl
C8H6Cl2O2
MFCD00000727
4-chlorophenoxyacetyl chloride, p-chlorophenoxyacetyl chloride, 2-4-chlorophenoxy acetyl chloride, 4-chlorophenoxy acetyl chloride, acetyl chloride, 4-chlorophenoxy, 4-chlorophenoxyacetylchloride, acmc-1aqz7, 4-chlorophenoxyacetyl chlroride, 4chlorophenoxy acetyl chloride, vrbvhqusaokvdh-uhfffaoysa
2-(4-chlorophenoxy)acetyl chloride

Specifications

4122-68-3
165°C
MFCD00000727
3265
VRBVHQUSAOKVDH-UHFFFAOYSA-N
2-(4-chlorophenoxy)acetyl chloride
77771
≥98.0% (T)
4-Chlorophenoxyacetyl Chloride
Yellow
C8H6Cl2O2
25 g
4-chlorophenoxyacetyl chloride, p-chlorophenoxyacetyl chloride, 2-4-chlorophenoxy acetyl chloride, 4-chlorophenoxy acetyl chloride, acetyl chloride, 4-chlorophenoxy, 4-chlorophenoxyacetylchloride, acmc-1aqz7, 4-chlorophenoxyacetyl chlroride, 4chlorophenoxy acetyl chloride, vrbvhqusaokvdh-uhfffaoysa
C1=CC(=CC=C1OCC(=O)Cl)Cl
205.034
205.03
Liquid
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'