missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorophenoxyacetonitrile, 98%

Manufacturer:  Alfa Aesar™ A1538906

 View more versions of this product

Catalog No. AAA1538906



Specifications

Specifications

4-Chlorophenoxyacetonitrile
45°C to 49°C
3598-13-8
MFCD00031535
2442163
YUGDKEWUYZXXRU-UHFFFAOYSA-N
2-(4-chlorophenoxy)acetonitrile
306444
98%
149°C to 150°C (15mmHg)
5g
C8H6ClNO
UN3439
2-4-chlorophenoxy acetonitrile, 4-chlorophenoxyacetonitrile, 4-chlorophenoxy acetonitrile, 4-chloro-phenoxy-acetonitrile, p-chlorophenoxyacetonitrile, 1-chloro-4-cyanomethoxy benzene, acetonitrile, 4-chlorophenoxy, 4-chloro-o-cyanomethylphenol, 2-4-chlorophenoxy ethanenitrile, acmc-20aopp
C1=CC(=CC=C1OCC#N)Cl
167.592
167.6

Chemical Identifiers

3598-13-8
167.592
YUGDKEWUYZXXRU-UHFFFAOYSA-N
306444
C1=CC(=CC=C1OCC#N)Cl
C8H6ClNO
MFCD00031535
2-4-chlorophenoxy acetonitrile, 4-chlorophenoxyacetonitrile, 4-chlorophenoxy acetonitrile, 4-chloro-phenoxy-acetonitrile, p-chlorophenoxyacetonitrile, 1-chloro-4-cyanomethoxy benzene, acetonitrile, 4-chlorophenoxy, 4-chloro-o-cyanomethylphenol, 2-4-chlorophenoxy ethanenitrile, acmc-20aopp
2-(4-chlorophenoxy)acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents