Learn More
4-chlorophenol, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 181000025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Chlorophenol | |
99.0 | |
1.2600g/mL | |
102°C | |
99% min. (GC) | |
C6H5ClO | |
MFCD00002318 | |
06,186 | |
15,2157 | |
Solubility in water: 2.7g/100 mL (20°C). Other solubilities: soluble in alcohol,ether,aromatic solvents,acetone,chloroform,glycerin and sodium hydroxide | |
C1=CC(=CC=C1O)Cl | |
128.56 | |
4.99 mPa.s (50°C) | |
128.56 |
106-48-9 | |
100.0 | |
220.0°C | |
Authentic | |
Glass bottle | |
ClC6H4OH | |
2.5kg | |
1.26 | |
p-chlorophenol, parachlorophenol, phenol, 4-chloro, 4-hydroxychlorobenzene, phenol, p-chloro, applied 3-78, 4-monochlorophenol, p-chlorophenic acid, p-chlorfenol, 4-chloro-1-hydroxybenzene | |
WXNZTHHGJRFXKQ-UHFFFAOYSA-N | |
4-chlorophenol | |
4684 | |
CHEBI:28078 | |
99+% |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath
GHS Signal Word: Warning
EINECSNumber : 203-402-6
RUO â Research Use Only