missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorophenol, 99%

Manufacturer:  Alfa Aesar™ A1560230

 View more versions of this product

Catalog No. AAA1560230



Description

Description

Applications
It is used as main raw material of compounding for medicine, dye, plastic and other industries. 4-Chlorophenol was used in visible-light-induced degradation of 4-chlorophenol in aqueous suspension of pure TiO2.

Solubility
Soluble in water (27g/L).

Notes
Air sensitive. Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

4-Chlorophenol
Phenol-like
39°C to 43°C
106-48-9
MFCD00002318
507004
p-chlorophenol, parachlorophenol, phenol, 4-chloro, 4-hydroxychlorobenzene, phenol, p-chloro, applied 3-78, 4-monochlorophenol, p-chlorophenic acid, p-chlorfenol, 4-chloro-1-hydroxybenzene
C1=CC(=CC=C1O)Cl
128.555
CHEBI:28078
99%
115°C (239°F)
217°C to 220°C
250g
C6H5ClO
UN2020
14,2154
WXNZTHHGJRFXKQ-UHFFFAOYSA-N
4-chlorophenol
4684
128.56

Chemical Identifiers

106-48-9
128.555
WXNZTHHGJRFXKQ-UHFFFAOYSA-N
4684
4-chlorophenol
C6H5ClO
MFCD00002318
p-chlorophenol, parachlorophenol, phenol, 4-chloro, 4-hydroxychlorobenzene, phenol, p-chloro, applied 3-78, 4-monochlorophenol, p-chlorophenic acid, p-chlorfenol, 4-chloro-1-hydroxybenzene
CHEBI:28078
C1=CC(=CC=C1O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P273-P302+P352
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID

RTECSNumber : SK2800000

TSCA : TSCA

Documents