Please login to your online account to display your discounted pricing

4-Chlorophenol 98%, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  109820025

 View more versions of this product

Catalog No. AC109820025

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Solid
Assay ≥97.5%
Formula Weight 128.56
Quantity 2.5L
Color White to pale yellow
EINECS Number 203-402-6
RTECS Number SK2800000
TSCA TSCA
Beilstein 06, 186
Merck Index 15, 2157
CAS Max % 100.0
Water 0.3% max.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Assay Percent Range 98%
CAS 106-48-9
MDL Number MFCD00002318
Chemical Name or Material 4-Chlorophenol
Linear Formula ClC6H4OH
Name Note 98%
Molecular Formula C6H5ClO
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 4-Chlorophenol
Name Note 98%
CAS 106-48-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H5ClO
Linear Formula ClC6H4OH
Formula Weight 128.56
MDL Number MFCD00002318
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

SDS & Safety Documents