Please login to your online account to display your discounted pricing

4-Chlorophenethylalcohol, 99%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  179580500

 View more versions of this product

Catalog No. AC179580500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 156.61
Physical Form Liquid
Quantity 50g
Color Clear colorless to slightly yellow
EINECS Number 217-506-4
CAS Max % 100.0
Chemical Name or Material 4-Chlorophenethylalcohol
Refractive Index 1.5470 to 1.5490 (20°C, 589nm)
Linear Formula ClC6H4CH2CH2OH
CAS 1875-88-3
MDL Number MFCD00002899
Molecular Formula C8H9ClO
Assay Percent Range 99%
Name Note 99%
Packaging Glass bottle
CAS Min % 98.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Chlorophenethylalcohol
Name Note 99%
CAS 1875-88-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H9ClO
Linear Formula ClC6H4CH2CH2OH
Formula Weight 156.61
MDL Number MFCD00002899