Please login to your online account to display your discounted pricing

4-Chlorophenethylalcohol, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chlorophenethylalcohol
Name Note 99%
CAS 1875-88-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H9ClO
Linear Formula ClC6H4CH2CH2OH
Formula Weight 156.61
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC179580500

  SDS
thermo fisher scientific
179580500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 156.61
Physical Form Liquid
Quantity 50g
Color Clear colorless to slightly yellow
EINECS Number 217-506-4
CAS Max % 100.0
Chemical Name or Material 4-Chlorophenethylalcohol
Refractive Index 1.5470 to 1.5490 (20°C, 589nm)
Linear Formula ClC6H4CH2CH2OH
CAS 1875-88-3
MDL Number MFCD00002899
Molecular Formula C8H9ClO
Assay Percent Range 99%
Name Note 99%
Packaging Glass bottle
CAS Min % 98.5
Infrared Spectrum Authentic