missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloronicotinic acid, 96%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC441720010
Specifications
4-Chloronicotinic acid | |
170°C to 171°C | |
10177-29-4 | |
C6H4ClNO2 | |
IMRGVWZLCZERSQ-UHFFFAOYSA-N | |
4-chloropyridine-3-carboxylic acid | |
818229 | |
96% |
Glass Bottle | |
1g | |
95% min. (HPLC) | |
3-carboxy-4-chloropyridine, 3-pyridinecarboxylic acid, 4-chloro, 4-chlornicotinsaure, 4-chloro-3-pyridinecarboxylic acid, 4-chloro-nicotinic acid, 4-chloronicotinic acid, 4-chloronicotinicacid, 4-chloropyridine-3-carboxyl, acmc-1br3v, pubchem12989 | |
C1=CN=CC(=C1Cl)C(=O)O | |
157.56 | |
157.56 |
Chemical Identifiers
10177-29-4 | |
157.56 | |
3-carboxy-4-chloropyridine, 3-pyridinecarboxylic acid, 4-chloro, 4-chlornicotinsaure, 4-chloro-3-pyridinecarboxylic acid, 4-chloro-nicotinic acid, 4-chloronicotinic acid, 4-chloronicotinicacid, 4-chloropyridine-3-carboxyl, acmc-1br3v, pubchem12989 | |
4-chloropyridine-3-carboxylic acid |
C6H4ClNO2 | |
IMRGVWZLCZERSQ-UHFFFAOYSA-N | |
818229 | |
C1=CN=CC(=C1Cl)C(=O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning