Please login to your online account to display your discounted pricing

4-Chloromethylbenzonitrile 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC391150100

 View more versions of this product

Catalog No. AC391150100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 151.59
Physical Form Crystalline Powder
Quantity 10g
Color Off-white to pale yellow
Melting Point 77° to 80°C
CAS Max % 100.0
Linear Formula ClCH2C6H4CN
Infrared Spectrum Authentic
Molecular Formula C8H6ClN
Chemical Name or Material 4-Chloromethylbenzonitrile
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Contact with acids liberates very toxic gas.
MDL Number MFCD00019761
CAS Min % 97.5
Packaging Glass bottle
CAS 874-86-2
Assay Percent Range 98%
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Chloromethylbenzonitrile
CAS 874-86-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H6ClN
Linear Formula ClCH2C6H4CN
Formula Weight 151.59
MDL Number MFCD00019761
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Contact with acids liberates very toxic gas.

SDS & Safety Documents