Learn More
4-(Chloromethyl)pyridine hydrochloride, 98%, Thermo Scientific Chemicals
$61.49 - $521.47
Chemical Identifiers
CAS | 1822-51-1 |
---|---|
Molecular Formula | C6H7Cl2N |
Molecular Weight (g/mol) | 164.029 |
MDL Number | MFCD00012826 |
InChI Key | ZDHKVKPZQKYREU-UHFFFAOYSA-N |
Synonym | 4-chloromethyl pyridine hydrochloride, 4-picolyl chloride hydrochloride, 4-chloromethyl pyridinium chloride, 4-chloromethylpyridine hydrochloride, 4-chloromethyl-pyridine hydrochloride, ccris 1728, isonicotinyl chloride hydrochloride, pyridine, 4-chloromethyl-, hydrochloride, 4-chloromethyl pyridine hcl, 4-chloromethyl pyridine, chloride |
PubChem CID | 74570 |
IUPAC Name | 4-(chloromethyl)pyridine;hydrochloride |
SMILES | C1=CN=CC=C1CCl.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1285906
|
Thermo Scientific Chemicals
A1285906 |
5 g |
Each for $61.49
|
|
|||||
AAA1285914
|
Thermo Scientific Chemicals
A1285914 |
25 g |
Each for $159.38
|
|
|||||
AAA1285922
|
Thermo Scientific Chemicals
A1285922 |
100 g |
Each for $521.47
|
|
|||||
Description
It finds its application as a reagent for protection of carboxyl termini of peptides as 4-picolyl esters, providing a polar 'handle' which aids in separation and purification of the peptide. The group can be introduced in the presence of a base, e.g. tetramethylguanidine, is stable to acid-catalyzed removal of Cbz protecting group and can be removed by base, Na in liquid ammonia or catalytic hydrogenolysis. It is also applied in conjunction with Amberlyst™15 resin in peptide synthesis. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application as a reagent for protection of carboxyl termini of peptides as 4-picolyl esters, providing a polar ′handle′ which aids in separation and purification of the peptide. The group can be introduced in the presence of a base, e.g. tetramethylguanidine, is stable to acid-catalyzed removal of Cbz protecting group and can be removed by base, Na in liquid ammonia or catalytic hydrogenolysis. It is also applied in conjunction with Amberlyst™15 resin in peptide synthesis. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in water (750 mg/ml at 20°C), soluble in 1N HCl (2.5%), DMSO, and methanol.
Notes
Hygroscopic. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
1822-51-1 | |
164.029 | |
ZDHKVKPZQKYREU-UHFFFAOYSA-N | |
74570 | |
C1=CN=CC=C1CCl.Cl |
C6H7Cl2N | |
MFCD00012826 | |
4-chloromethyl pyridine hydrochloride, 4-picolyl chloride hydrochloride, 4-chloromethyl pyridinium chloride, 4-chloromethylpyridine hydrochloride, 4-chloromethyl-pyridine hydrochloride, ccris 1728, isonicotinyl chloride hydrochloride, pyridine, 4-chloromethyl-, hydrochloride, 4-chloromethyl pyridine hcl, 4-chloromethyl pyridine, chloride | |
4-(chloromethyl)pyridine;hydrochloride |
Specifications
1822-51-1 | |
C6H7Cl2N | |
5 g | |
3689154 | |
4-chloromethyl pyridine hydrochloride, 4-picolyl chloride hydrochloride, 4-chloromethyl pyridinium chloride, 4-chloromethylpyridine hydrochloride, 4-chloromethyl-pyridine hydrochloride, ccris 1728, isonicotinyl chloride hydrochloride, pyridine, 4-chloromethyl-, hydrochloride, 4-chloromethyl pyridine hcl, 4-chloromethyl pyridine, chloride | |
ZDHKVKPZQKYREU-UHFFFAOYSA-N | |
4-(chloromethyl)pyridine;hydrochloride | |
74570 | |
98% |
166°C to 171°C | |
MFCD00012826 | |
UN3261 | |
Hygroscopic | |
Soluble in water (750mg/ml at 20°C),soluble in 1N HCl (2.5%),DMSO,and methanol. | |
C1=CN=CC=C1CCl.Cl | |
164.029 | |
164.04 | |
4-(Chloromethyl)pyridine hydrochloride |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 217-350-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only