missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloromethyl-N,N-diisopropylbenzamide, 97%

$220.53 - $703.20

Chemical Identifiers

CAS 1375930-97-4
Molecular Formula C14H20ClNO
Molecular Weight (g/mol) 253.77
MDL Number MFCD22683130
InChI Key IWHDTZURUKJHFU-UHFFFAOYSA-N
Synonym 4-chloromethyl-n,n-diisopropylbenzamide, 4-chloromethyl-n,n-diisopropylbenzamide
PubChem CID 98000579
IUPAC Name 4-(chloromethyl)-N,N-di(propan-2-yl)benzamide
SMILES CC(C)N(C(C)C)C(=O)C1=CC=C(C=C1)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60486MD View Documents Alfa Aesar™
H60486MD
250mg Each for $220.53
 
AAH6048603 View Documents Alfa Aesar™
H6048603
1g Each for $703.20
 
Specifications

Chemical Identifiers

1375930-97-4
253.77
IWHDTZURUKJHFU-UHFFFAOYSA-N
98000579
CC(C)N(C(C)C)C(=O)C1=CC=C(C=C1)CCl
C14H20ClNO
MFCD22683130
4-chloromethyl-n,n-diisopropylbenzamide, 4-chloromethyl-n,n-diisopropylbenzamide
4-(chloromethyl)-N,N-di(propan-2-yl)benzamide

Specifications

250mg
C14H20ClNO
22574301
IWHDTZURUKJHFU-UHFFFAOYSA-N
4-(chloromethyl)-N,N-di(propan-2-yl)benzamide
98000579
97%
1375930-97-4
MFCD22683130
4-chloromethyl-n,n-diisopropylbenzamide, 4-chloromethyl-n,n-diisopropylbenzamide
CC(C)N(C(C)C)C(=O)C1=CC=C(C=C1)CCl
253.77
253.77
4-Chloromethyl-N,N-diisopropylbenzamide