missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloromethyl-N-(4-fluorobenzyl)benzamide, 97%

Chemical Identifiers

CAS 1094362-88-5
Molecular Formula C15H13ClFNO
Molecular Weight (g/mol) 277.723
MDL Number MFCD12568403
InChI Key IWRWVLGYZZVJJR-UHFFFAOYSA-N
Synonym 4-chloromethyl-n-4-fluorobenzyl benzamide, 4-chloromethyl-n-4-fluorophenyl methyl benzamide
PubChem CID 43156288
IUPAC Name 4-(chloromethyl)-N-[(4-fluorophenyl)methyl]benzamide
SMILES C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)CCl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60128MD
View Documents
Alfa Aesar™
H60128MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6012803
View Documents
Alfa Aesar™
H6012803
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1094362-88-5
277.723
IWRWVLGYZZVJJR-UHFFFAOYSA-N
43156288
C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)CCl)F
C15H13ClFNO
MFCD12568403
4-chloromethyl-n-4-fluorobenzyl benzamide, 4-chloromethyl-n-4-fluorophenyl methyl benzamide
4-(chloromethyl)-N-[(4-fluorophenyl)methyl]benzamide

Specifications

250mg
C15H13ClFNO
4-chloromethyl-n-4-fluorobenzyl benzamide, 4-chloromethyl-n-4-fluorophenyl methyl benzamide
C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)CCl)F
277.723
277.72
4-Chloromethyl-N-(4-fluorobenzyl)benzamide
1094362-88-5
MFCD12568403
IWRWVLGYZZVJJR-UHFFFAOYSA-N
4-(chloromethyl)-N-[(4-fluorophenyl)methyl]benzamide
43156288
97%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures