missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloromethyl-N-(3,5-difluorophenyl)benzamide, 97%

Chemical Identifiers

CAS 1094362-94-3
Molecular Formula C14H10ClF2NO
Molecular Weight (g/mol) 281.687
MDL Number MFCD12540820
InChI Key AQOFMGRCNVYGFX-UHFFFAOYSA-N
Synonym 4-chloromethyl-n-3,5-difluorophenyl benzamide, 4-chloromethyl-n-3,5-difluorophenyl benzamide
PubChem CID 43156300
IUPAC Name 4-(chloromethyl)-N-(3,5-difluorophenyl)benzamide
SMILES C1=CC(=CC=C1CCl)C(=O)NC2=CC(=CC(=C2)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60628MD View Documents Alfa Aesar™
H60628MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6062803 View Documents Alfa Aesar™
H6062803
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1094362-94-3
281.687
AQOFMGRCNVYGFX-UHFFFAOYSA-N
43156300
C1=CC(=CC=C1CCl)C(=O)NC2=CC(=CC(=C2)F)F
C14H10ClF2NO
MFCD12540820
4-chloromethyl-n-3,5-difluorophenyl benzamide, 4-chloromethyl-n-3,5-difluorophenyl benzamide
4-(chloromethyl)-N-(3,5-difluorophenyl)benzamide

Specifications

250mg
C14H10ClF2NO
4-chloromethyl-n-3,5-difluorophenyl benzamide, 4-chloromethyl-n-3,5-difluorophenyl benzamide
C1=CC(=CC=C1CCl)C(=O)NC2=CC(=CC(=C2)F)F
281.687
281.69
4-Chloromethyl-N-(3,5-difluorophenyl)benzamide
1094362-94-3
MFCD12540820
AQOFMGRCNVYGFX-UHFFFAOYSA-N
4-(chloromethyl)-N-(3,5-difluorophenyl)benzamide
43156300
97%