missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloromethyl-N-(2-ethylhexyl)benzamide, 97%

Chemical Identifiers

CAS 1293014-05-7
Molecular Formula C16H24ClNO
Molecular Weight (g/mol) 281.824
MDL Number MFCD13317402
InChI Key JLQFBEBBEBVSIV-UHFFFAOYSA-N
Synonym 4-chloromethyl-n-2-ethylhexyl benzamide, 4-chloromethyl-n-2-ethylhexyl benzamide
PubChem CID 61251738
IUPAC Name 4-(chloromethyl)-N-(2-ethylhexyl)benzamide
SMILES CCCCC(CC)CNC(=O)C1=CC=C(C=C1)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60715MD View Documents Alfa Aesar™
H60715MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6071503 View Documents Alfa Aesar™
H6071503
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1293014-05-7
281.824
JLQFBEBBEBVSIV-UHFFFAOYSA-N
61251738
CCCCC(CC)CNC(=O)C1=CC=C(C=C1)CCl
C16H24ClNO
MFCD13317402
4-chloromethyl-n-2-ethylhexyl benzamide, 4-chloromethyl-n-2-ethylhexyl benzamide
4-(chloromethyl)-N-(2-ethylhexyl)benzamide

Specifications

250mg
C16H24ClNO
4-chloromethyl-n-2-ethylhexyl benzamide, 4-chloromethyl-n-2-ethylhexyl benzamide
CCCCC(CC)CNC(=O)C1=CC=C(C=C1)CCl
281.824
281.82
4-Chloromethyl-N-(2-ethylhexyl)benzamide
1293014-05-7
MFCD13317402
JLQFBEBBEBVSIV-UHFFFAOYSA-N
4-(chloromethyl)-N-(2-ethylhexyl)benzamide
61251738
97%