Please login to your online account to display your discounted pricing

4-(chloromethyl)benzoyl chloride, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  314440050

 View more versions of this product

Catalog No. AC314440050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 189.04
Physical Form Low Melting Crystalline Mass
Quantity 5g
Color White to light beige
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula ClCHC6H4COCl
Chemical Name or Material 4-(chloromethyl)benzoyl chloride
Assay Percent Range 97%
Molecular Formula C8H6Cl2O
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00053224
CAS 876-08-4
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-(chloromethyl)benzoyl chloride
CAS 876-08-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H6Cl2O
Linear Formula ClCHC6H4COCl
Formula Weight 189.04
MDL Number MFCD00053224
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents