missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Chloromethyl)benzoic acid, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2227514

 View more versions of this product

Catalog No. AAB2227514



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(Chloromethyl)benzoic acid
199°C to 202°C
MFCD00002568
UN3261
4-chloromethyl benzoic acid, alpha-chloro-p-toluic acid, benzoic acid, 4-chloromethyl, 4-carboxybenzyl chloride, 4-chloromethyl-benzoic acid, alpha-chloro-p-toluylic acid, .alpha.-chloro-p-toluylic acid, pubchem15446, alpha-chloroparatoluic acid
C1=CC(=CC=C1CCl)C(=O)O
170.592
170.6
1642-81-5
C8H7ClO2
25 g
1907970
OITNBJHJJGMFBN-UHFFFAOYSA-N
4-(chloromethyl)benzoic acid
74234
96%

Chemical Identifiers

1642-81-5
170.592
OITNBJHJJGMFBN-UHFFFAOYSA-N
74234
C1=CC(=CC=C1CCl)C(=O)O
C8H7ClO2
MFCD00002568
4-chloromethyl benzoic acid, alpha-chloro-p-toluic acid, benzoic acid, 4-chloromethyl, 4-carboxybenzyl chloride, 4-chloromethyl-benzoic acid, alpha-chloro-p-toluylic acid, .alpha.-chloro-p-toluylic acid, pubchem15446, alpha-chloroparatoluic acid
4-(chloromethyl)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 216-697-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only