missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 4-Chloromethyl-5-methyl-2-oxo-1,3-dioxole, 97%
$29.54 - $273.20
Chemical Identifiers
CAS | 80841-78-7 |
---|---|
Molecular Formula | C5H5ClO3 |
Molecular Weight (g/mol) | 148.542 |
MDL Number | MFCD07787494 |
InChI Key | QCLFSYYUWPUWQR-UHFFFAOYSA-N |
Synonym | 1,3-dioxol-2-one, 4-chloromethyl-5-methyl, 4-chloromethyl-5-methyl-1,3-dioxol-2-one, 4-chloromethyl-5-methyl-1,3-dioxolen-2-one, 4-chloromethyl-5-methyl-2h-1,3-dioxol-2-one, 4-chloromethyl-5methyl-1,3-dioxol-2-one, 4-choromethyl-5-methyl-2-oxo-1,3-dioxole, 4-cloromethyl-5-methyl-1,3-dioxol-2-one, acmc-209pk7, ksc490e4r, pubchem10542 |
PubChem CID | 9855518 |
IUPAC Name | 4-(chloromethyl)-5-methyl-1,3-dioxol-2-one |
SMILES | CC1=C(OC(=O)O1)CCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH55735MD
|
Alfa Aesar™
H55735MD |
250mg |
Each for $29.54
|
|
AAH5573503
|
Alfa Aesar™
H5573503 |
1g |
Each for $80.84
|
|
AAH5573506
|
Alfa Aesar™
H5573506 |
5g |
Each for $273.20
|
|
Chemical Identifiers
80841-78-7 | |
148.542 | |
QCLFSYYUWPUWQR-UHFFFAOYSA-N | |
9855518 | |
CC1=C(OC(=O)O1)CCl |
C5H5ClO3 | |
MFCD07787494 | |
1,3-dioxol-2-one, 4-chloromethyl-5-methyl, 4-chloromethyl-5-methyl-1,3-dioxol-2-one, 4-chloromethyl-5-methyl-1,3-dioxolen-2-one, 4-chloromethyl-5-methyl-2h-1,3-dioxol-2-one, 4-chloromethyl-5methyl-1,3-dioxol-2-one, 4-choromethyl-5-methyl-2-oxo-1,3-dioxole, 4-cloromethyl-5-methyl-1,3-dioxol-2-one, acmc-209pk7, ksc490e4r, pubchem10542 | |
4-(chloromethyl)-5-methyl-1,3-dioxol-2-one |
Specifications
129°C (264°F) | |
250mg | |
C5H5ClO3 | |
4798926 | |
QCLFSYYUWPUWQR-UHFFFAOYSA-N | |
4-(chloromethyl)-5-methyl-1,3-dioxol-2-one | |
9855518 | |
97% |
>90°C (2mmHg) | |
80841-78-7 | |
MFCD07787494 | |
1,3-dioxol-2-one, 4-chloromethyl-5-methyl, 4-chloromethyl-5-methyl-1,3-dioxol-2-one, 4-chloromethyl-5-methyl-1,3-dioxolen-2-one, 4-chloromethyl-5-methyl-2h-1,3-dioxol-2-one, 4-chloromethyl-5methyl-1,3-dioxol-2-one, 4-choromethyl-5-methyl-2-oxo-1,3-dioxole, 4-cloromethyl-5-methyl-1,3-dioxol-2-one, acmc-209pk7, ksc490e4r, pubchem10542 | |
CC1=C(OC(=O)O1)CCl | |
148.542 | |
148.55 | |
4-Chloromethyl-5-methyl-2-oxo-1,3-dioxole |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures