missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloromethyl-5-methyl-2-oxo-1,3-dioxole, 97%

Manufacturer:  Alfa Aesar™ H55735MD

 View more versions of this product

Catalog No. AAH55735MD



Specifications

Specifications

4-Chloromethyl-5-methyl-2-oxo-1,3-dioxole
>90°C (2mmHg)
80841-78-7
MFCD07787494
1,3-dioxol-2-one, 4-chloromethyl-5-methyl
CC1=C(OC(=O)O1)CCl
148.542
148.55
129°C (264°F)
250mg
C5H5ClO3
4798926
QCLFSYYUWPUWQR-UHFFFAOYSA-N
4-(chloromethyl)-5-methyl-1,3-dioxol-2-one
9855518
97%

Chemical Identifiers

80841-78-7
148.542
QCLFSYYUWPUWQR-UHFFFAOYSA-N
9855518
CC1=C(OC(=O)O1)CCl
C5H5ClO3
MFCD07787494
1,3-dioxol-2-one, 4-chloromethyl-5-methyl
4-(chloromethyl)-5-methyl-1,3-dioxol-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents