Please login to your online account to display your discounted pricing

4-(Chloromethyl)-3,5-dimethylisoxazole, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(Chloromethyl)-3, 5-dimethylisoxazole
Assay Percent Range 97%
Molecular Formula C6H8ClNO
Formula Weight 145.59
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

JFD00628DA

  SDS
fisher scientific company
JFD00628DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 145.59
Quantity 1g
Chemical Name or Material 4-(Chloromethyl)-3, 5-dimethylisoxazole
Assay Percent Range 97%
Molecular Formula C6H8ClNO
CAS 19788-37-5
Packaging Amber Glass Bottle