Please login to your online account to display your discounted pricing

4-(Chloromethyl)-3,5-dimethylisoxazole, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  JFD00628DA

 View more versions of this product

Catalog No. JFD00628DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 145.59
Quantity 1g
Chemical Name or Material 4-(Chloromethyl)-3, 5-dimethylisoxazole
Assay Percent Range 97%
Molecular Formula C6H8ClNO
CAS 19788-37-5
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4-(Chloromethyl)-3, 5-dimethylisoxazole
Assay Percent Range 97%
Molecular Formula C6H8ClNO
Formula Weight 145.59