missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloromethyl-3,5-dimethylisoxazole, 97%

$48.12 - $564.42

Chemical Identifiers

CAS 19788-37-5
Molecular Formula C6H8ClNO
Molecular Weight (g/mol) 145.586
MDL Number MFCD00003154
InChI Key NIFAUKBQIAURIM-UHFFFAOYSA-N
Synonym 4-chloromethyl-3,5-dimethylisoxazole, 4-chloromethyl-3,5-dimethylisoxazole, isoxazole, 4-chloromethyl-3,5-dimethyl, 4-chloromethyl-3,5-dimethyl-1,2-oxazole, 4-chloromethyl-3,5-dimethyl-isoxazole, 3,5-dimethyl-4-chloromethylisoxazole, 4-chloromethyl-3,5-dimethyl isoxazole, pubchem8667, acmc-1c8qy, akos bc-3064
PubChem CID 88246
IUPAC Name 4-(chloromethyl)-3,5-dimethyl-1,2-oxazole
SMILES CC1=C(C(=NO1)C)CCl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0051503 View Documents Alfa Aesar™
L0051503
1g Each for $48.12
 
AAL0051506 View Documents Alfa Aesar™
L0051506
5g Each for $153.12
 
AAL0051514 View Documents Alfa Aesar™
L0051514
25g Each for $564.42
 
Specifications

Chemical Identifiers

19788-37-5
145.586
NIFAUKBQIAURIM-UHFFFAOYSA-N
88246
CC1=C(C(=NO1)C)CCl
C6H8ClNO
MFCD00003154
4-chloromethyl-3,5-dimethylisoxazole, 4-chloromethyl-3,5-dimethylisoxazole, isoxazole, 4-chloromethyl-3,5-dimethyl, 4-chloromethyl-3,5-dimethyl-1,2-oxazole, 4-chloromethyl-3,5-dimethyl-isoxazole, 3,5-dimethyl-4-chloromethylisoxazole, 4-chloromethyl-3,5-dimethyl isoxazole, pubchem8667, acmc-1c8qy, akos bc-3064
4-(chloromethyl)-3,5-dimethyl-1,2-oxazole

Specifications

95°C (203°F)
1g
C6H8ClNO
110780
NIFAUKBQIAURIM-UHFFFAOYSA-N
4-(chloromethyl)-3,5-dimethyl-1,2-oxazole
88246
97%
88°C to 90°C (7mmHg)
19788-37-5
MFCD00003154
4-chloromethyl-3,5-dimethylisoxazole, 4-chloromethyl-3,5-dimethylisoxazole, isoxazole, 4-chloromethyl-3,5-dimethyl, 4-chloromethyl-3,5-dimethyl-1,2-oxazole, 4-chloromethyl-3,5-dimethyl-isoxazole, 3,5-dimethyl-4-chloromethylisoxazole, 4-chloromethyl-3,5-dimethyl isoxazole, pubchem8667, acmc-1c8qy, akos bc-3064
CC1=C(C(=NO1)C)CCl
145.586
145.59
4-Chloromethyl-3,5-dimethylisoxazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA