Please login to your online account to display your discounted pricing

4-(chloromethyl)-3,5-dimethylisoxazole, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(chloromethyl)-3, 5-dimethylisoxazole
CAS 19788-37-5
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C6H8ClNO
Formula Weight 145.59
MDL Number MFCD00003154
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC355390010

  SDS
thermo fisher scientific
355390010
Glass bottle 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Liquid
Formula Weight 145.59
Quantity 1mL
Color Clear yellow
Beilstein 27, 19
CAS Min % 95.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96%
Molecular Formula C6H8ClNO
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00003154
Packaging Glass bottle
Refractive Index 1.4860 to 1.4880 (20°C, 589nm)
Chemical Name or Material 4-(chloromethyl)-3, 5-dimethylisoxazole
CAS 19788-37-5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.