Please login to your online account to display your discounted pricing

4-(chloromethyl)-3,5-dimethylisoxazole, 96%, ACROS Organics™


Quantity 1mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  355390010

 View more versions of this product

Catalog No. AC355390010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid
Formula Weight 145.59
Quantity 1mL
Color Clear yellow
Beilstein 27, 19
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C6H8ClNO
MDL Number MFCD00003154
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 4-(chloromethyl)-3, 5-dimethylisoxazole
Refractive Index 1.4860 to 1.4880 (20°C, 589nm)
CAS 19788-37-5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 4-(chloromethyl)-3, 5-dimethylisoxazole
CAS 19788-37-5
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C6H8ClNO
Formula Weight 145.59
MDL Number MFCD00003154
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents