Please login to your online account to display your discounted pricing

4-(Chloromethyl)-2-methyl-1,3-thiazole hydrochloride , Tech., Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CD09497CB

 View more versions of this product

Catalog No. CD09497CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 184.09
Purity Grade Technical Grade
Quantity 250mg
Assay Tech.
CAS 77470-53-2
Molecular Formula C5H6ClNS·ClH
Packaging Amber glass bottle
Chemical Name or Material 4-(Chloromethyl)-2-methyl-1, 3-thiazole hydrochloride

Specifications

Chemical Name or Material 4-(Chloromethyl)-2-methyl-1, 3-thiazole hydrochloride
Assay Tech.
Molecular Formula C5H6ClNS·ClH
Formula Weight 184.09