missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Chloromethyl)-2,5-dimethyl-1,3-oxazole, ≥97%, Maybridge

$411.00 - $411.00

Chemical Identifiers

CAS 777823-76-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H8ClNO
Linear Formula Yes
Formula Weight 145.59
MDL Number MFCD03923825
Chemical Name or Material 4-(Chloromethyl)-2, 5-dimethyl-1, 3-oxazole
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC26824DA SDS Fisher Scientific Uk Ltd
CC26824DA
Amber Glass Bottle 1g Each for $411.00
Specifications Description & Specifications

Chemical Identifiers

777823-76-4
100.0
C6H8ClNO
145.59
4-(Chloromethyl)-2, 5-dimethyl-1, 3-oxazole
96.5
97%
Yes
MFCD03923825
97%

Specifications

777823-76-4
100.0
97%
C6H8ClNO
145.59
4-(Chloromethyl)-2, 5-dimethyl-1, 3-oxazole
96.5
Amber Glass Bottle
1g
Yes
MFCD03923825
97%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.