Please login to your online account to display your discounted pricing

4-(Chloromethyl)-2,5-dimethyl-1,3-oxazole, ≥97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific uk ltd  CC26824DA

 View more versions of this product

Catalog No. CC26824DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 145.59
Quantity 1g
Assay 97%
CAS Max % 100.0
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Chemical Name or Material 4-(Chloromethyl)-2, 5-dimethyl-1, 3-oxazole
Assay Percent Range 97%
MDL Number MFCD03923825
Molecular Formula C6H8ClNO
CAS Min % 96.5
Linear Formula Yes
CAS 777823-76-4

Specifications

Chemical Name or Material 4-(Chloromethyl)-2, 5-dimethyl-1, 3-oxazole
CAS 777823-76-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H8ClNO
Linear Formula Yes
Formula Weight 145.59
MDL Number MFCD03923825
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents