Please login to your online account to display your discounted pricing

4-(Chloromethyl)-2-(3-chlorophenyl)-1,3-thiazole hydrochloride, 97+%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(Chloromethyl)-2-(3-chlorophenyl)-1, 3-thiazole hydrochloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7Cl2NS·ClH
Formula Weight 280.61
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO01135CB

  SDS
fisher scientific company
MO01135CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 280.61
Quantity 250mg
Assay 97%
Chemical Name or Material 4-(Chloromethyl)-2-(3-chlorophenyl)-1, 3-thiazole hydrochloride
Assay Percent Range 97%
Molecular Formula C10H7Cl2NS·ClH
CAS 690632-83-8