Please login to your online account to display your discounted pricing

4-(Chloromethyl)-2-(3-chlorophenyl)-1,3-thiazole hydrochloride, 97+%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  MO01135CB

 View more versions of this product

Catalog No. MO01135CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 280.61
Quantity 250mg
Assay 97%
Chemical Name or Material 4-(Chloromethyl)-2-(3-chlorophenyl)-1, 3-thiazole hydrochloride
Assay Percent Range 97%
Molecular Formula C10H7Cl2NS·ClH
CAS 690632-83-8

Specifications

Chemical Name or Material 4-(Chloromethyl)-2-(3-chlorophenyl)-1, 3-thiazole hydrochloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7Cl2NS·ClH
Formula Weight 280.61