Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Chloromethyl)-1,2-diphenylethane, 98%

Chemical Identifiers

Chemical Name or Material 4-(Chloromethyl)-1,2-diphenylethane
CAS 80676-35-3
Molecular Formula C15H15Cl
Formula Weight 230.73
MDL Number MFCD02093918
UN Number UN1759
Synonym 4-(Chloromethyl)bibenzyl; 4-(Chloromethyl)dibenzyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2522203

 
alfa aesar™
B2522203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2522206

 
alfa aesar™
B2522206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2522214

 
alfa aesar™
B2522214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.73
Assay 98%
CAS 80676-35-3
UN Number UN1759
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 38° to 42°C
Chemical Name or Material 4-(Chloromethyl)-1,2-diphenylethane
Molecular Formula C15H15Cl
MDL Number MFCD02093918
Synonym 4-(Chloromethyl)bibenzyl; 4-(Chloromethyl)dibenzyl
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.