Please login to your online account to display your discounted pricing

4-Chloroindole, 99%, ACROS Organics™

$250.76 - $250.76

Chemical Identifiers

Chemical Name or Material 4-Chloroindole
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H6ClN
Formula Weight 151.6
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC123230010

  SDS
thermo fisher scientific
123230010
Glass bottle 1g Each for $250.76
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 151.6
Quantity 1g
Color Clear yellow
Density 1.2500g/mL
Packaging Glass bottle
Refractive Index 1.6270 to 1.6290 (20°C, 589nm)
CAS 25235-85-2
Additional Information May darken on storage
Infrared Spectrum Authentic
Chemical Name or Material 4-Chloroindole
Assay Percent Range 99%
Molecular Formula C8H6ClN