missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorocinnamic acid, 99%, predominantly trans, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 108941000

 View more versions of this product

Catalog No. AC108941000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Chlorocinnamic acid
Plastic bottle
246°C to 250°C
99%
98.5
99%
ClC6H4CH=CHCO2H
09, 594
GXLIFJYFGMHYDY-ZZXKWVIFSA-N
(E)-3-(4-chlorophenyl)prop-2-enoic acid
637797
182.61
≥98.5%
Authentic
White
100g
1615-02-7
100.0
C9H7ClO2
MFCD00004396
2-propenoic acid, 3-4-chlorophenyl, 2e-3-4-chlorophenyl prop-2-enoic acid, 3-4-chlorophenyl acrylic acid, 3-p-chlorophenyl acrylic acid, 4-chlorocinnamic acid, cinnamic acid, p-chloro, e-p-chlorocinnamic acid, p-chlorocinnamic acid, trans-4-chlorocinnamic acid, trans-p-chlorocinnamic acid
OC(=O)\C=C\C1=CC=C(Cl)C=C1
182.60
CHEBI:61116
Fine Crystalline Powder

Chemical Identifiers

1615-02-7
182.60
GXLIFJYFGMHYDY-ZZXKWVIFSA-N
637797
(E)-3-(4-chlorophenyl)prop-2-enoic acid
C9H7ClO2
MFCD00004396
2-propenoic acid, 3-4-chlorophenyl, 2e-3-4-chlorophenyl prop-2-enoic acid, 3-4-chlorophenyl acrylic acid, 3-p-chlorophenyl acrylic acid, 4-chlorocinnamic acid, cinnamic acid, p-chloro, e-p-chlorocinnamic acid, p-chlorocinnamic acid, trans-4-chlorocinnamic acid, trans-p-chlorocinnamic acid
CHEBI:61116
OC(=O)\C=C\C1=CC=C(Cl)C=C1
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 216-564-8

RTECSNumber : GD8490000

SDS Documents