missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorocinnamic acid, predominantly trans, 99%

Manufacturer:  Alfa Aesar™ A1530436

 View more versions of this product

Catalog No. AAA1530436



Specifications

Specifications

4-Chlorocinnamic acid
500g
1615-02-7
MFCD00004396
4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid
C1=CC(=CC=C1C=CC(=O)O)Cl
182.603
CHEBI:61116
99%
248°C to 253°C
predominantly trans
C9H7ClO2
1365308
GXLIFJYFGMHYDY-ZZXKWVIFSA-N
(E)-3-(4-chlorophenyl)prop-2-enoic acid
637797
182.61

Chemical Identifiers

1615-02-7
182.603
GXLIFJYFGMHYDY-ZZXKWVIFSA-N
637797
(E)-3-(4-chlorophenyl)prop-2-enoic acid
C9H7ClO2
MFCD00004396
4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid
CHEBI:61116
C1=CC(=CC=C1C=CC(=O)O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : GD8490000

Documents